MilliporeSigma™ Supelco™ Screw Top Mini Flask with Hole Cap and Septum
Heat resistant borosilicate glass flasks provide an inert seal when used with a hole cap and PTFE silicone septum (included) or a Mininert valve.
Supplier:  MilliporeSigma™ Supelco™ 64716U
 View more versions of this product
Catalog No. 111009783
Explore available promotions
Description
Description
Excellent for liquid-liquid extraction of trihalomethanes from drinking water. Also useful for sample collection, solution preparations, derivatization reactions, extractions, and many other applications.
Specifications
Specifications
Clear
25 mL
12/Pk.
Borosilicate Glass