Learn More
Ab Sciex Llc SDS-Gel MW Analysis Kit for use on the PA 800 Plus DFS Item
Supplier:  Danaher Corp 390953
Protein Chemistry Kit; ProteomeLab SDS-Gel MW Analysis Chemistries; Beckman Coulter; Sieving range 10kDa to 225kD; Inc. buffers, standards and washes; For quantification and determination of purity of proteins
Catalog No. NC9118142
Only null left
Explore available promotions