missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisher Science Education™ Seamless Cellulose Dialysis Tubing
Retains materials with 12,000 and higher molecular weight.
Supplier: Fisher Science Education™ S25645
Description
- Especially useful in analytical chemistry, biology, and pharmaceutical fields
- Highly permeable to water and low molecular weight compounds
- Contains glycerin, water and 1% sulfur
- Must be stored in airtight container or rehumidified before use
- Pore diameter: 4.8nm
This product is an Educational Grade Chemical — no C of A is available. ‡ Average values.
Specifications
Dialysis Tubing | |
15.9 mm | |
10 ft. | |
Glycerin, water and 1% sulfur | |
Useful in analytical chemistry, biology, and pharmaceutical fields |
Seamless Cellulose | |
0.62 in. | |
3 m | |
Each |