missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisher Science Education™ Seamless Cellulose Dialysis Tubing

Retains materials with 12,000 and higher molecular weight.

Supplier:  Fisher Science Education™ S25645B

 View more versions of this product

Catalog No. S25645B

Explore available promotions

Description

Description

  • Especially useful in analytical chemistry, biology, and pharmaceutical fields
  • Highly permeable to water and low molecular weight compounds
  • Contains glycerin, water and 1% sulfur
  • Must be stored in airtight container or rehumidified before use
  • Pore diameter: 4.8nm

This product is an Educational Grade Chemical — no C of A is available. ‡ Average values.

Specifications

Specifications

Dialysis Tubing
28.6 mm
10 ft.
Glycerin, water and 1% sulfur
Useful in analytical chemistry, biology, and pharmaceutical fields
Seamless Cellulose
1.13 in.
3 m
Each
SDS
Documents

Documents

Promotions

Promotions