missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sebacic acid, 98+%, Thermo Scientific Chemicals

$50.77 - $688.47

Chemical Identifiers

CAS 111-20-6
Molecular Formula C10H18O4
Molecular Weight (g/mol) 202.25
MDL Number MFCD00004440
InChI Key CXMXRPHRNRROMY-UHFFFAOYSA-N
Synonym sebacic acid, 1,8-octanedicarboxylic acid, 1,10-decanedioic acid, sebacic acids, decanedicarboxylic acid, n-decanedioic acid, sebacinsaure, acide sebacique, sebacate, sebacinsaeure
PubChem CID 5192
ChEBI CHEBI:41865
IUPAC Name decanedioic acid
SMILES C(CCCCC(=O)O)CCCC(=O)O
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1415822
View Documents
Thermo Scientific Chemicals
A1415822
100 g
Each for $50.77
 
AAA1415830
View Documents
Thermo Scientific Chemicals
A1415830
250 g
Each for $64.08
 
AAA141580B
View Documents
Thermo Scientific Chemicals
A141580B
1000 g
Each for $176.23
 
AAA141580I
View Documents
Thermo Scientific Chemicals
A141580I
5000 g
Each for $688.47
 
Description

Description

Sebacic acid is used as a monomer for nylon 610, plasticizers, lubricants, hydraulic fluids, cosmetics and candles. It is also used as intermediates in antiseptics, aromatics and painting materials. It is used in the synthesis of poly(glycerol sebacate) and novel bio-nylon. It finds application as adhesives, corrosion inhibitors and solvents. Its derivative, diethyl Sebacate is used in plastics manufacturing and engineering, as well as drug delivery procedures through the use of biodegradable encapsulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sebacic acid is used as a monomer for nylon 610, plasticizers, lubricants, hydraulic fluids, cosmetics and candles. It is also used as intermediates in antiseptics, aromatics and painting materials. It is used in the synthesis of poly(glycerol sebacate) and novel bio-nylon. It finds application as adhesives, corrosion inhibitors and solvents. Its derivative, diethyl Sebacate is used in plastics manufacturing and engineering, as well as drug delivery procedures through the use of biodegradable encapsulations.

Solubility
Slightly soluble in water.

Notes
Incompatible with bases, reducing agents and oxidizing agents.
Specifications

Chemical Identifiers

111-20-6
202.25
CXMXRPHRNRROMY-UHFFFAOYSA-N
5192
decanedioic acid
C10H18O4
MFCD00004440
sebacic acid, 1,8-octanedicarboxylic acid, 1,10-decanedioic acid, sebacic acids, decanedicarboxylic acid, n-decanedioic acid, sebacinsaure, acide sebacique, sebacate, sebacinsaeure
CHEBI:41865
C(CCCCC(=O)O)CCCC(=O)O

Specifications

111-20-6
1.271
220°C (428°F)
HO2C(CH2)8CO2H
100 g
14,8415
Slightly soluble in water.
C(CCCCC(=O)O)CCCC(=O)O
202.25
CHEBI:41865
≥98%
131°C to 134°C
295°C (100 mmHg)
C10H18O4
MFCD00004440
1210591
sebacic acid, 1,8-octanedicarboxylic acid, 1,10-decanedioic acid, sebacic acids, decanedicarboxylic acid, n-decanedioic acid, sebacinsaure, acide sebacique, sebacate, sebacinsaeure
CXMXRPHRNRROMY-UHFFFAOYSA-N
decanedioic acid
5192
202.25
Sebacic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 203-845-5

RTECSNumber : VS0875000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only