missing translation for 'onlineSavingsMsg'
Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits

Sebacoyl chloride, 92%, technical, Thermo Scientific Chemicals

Sebacoyl chloride, 92%, C10H16Cl2O2, CAS Number-111-19-3, decanedioic acid chloride, sebacoyl chloride, cp, octane-1,8-dicarbonyl chloride, sebacyl chloride, sebacoyl dichloride, sebacic acid dichloride, sebacoyl chloride, sebacoylchloride, acmc-1byrk, decanedioyl chloride, 500g, 90% min. (GC)

Supplier:  Thermo Scientific Chemicals 294900500

 Afficher plus de versions de ce produit

Numéro de catalogue. AC294900500


Découvrez plus d'offres spéciales

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Spécifications

Sebacoyl chloride
-2.5°C
1.1210g/mL
>110°C
90% min. (GC)
C10H16Cl2O2
ClCO(CH2)8COCl
50 g
1.121
Solubility in water: may decompose. Other solubilities: soluble in hydrocarbons and ethers
C(CCCCC(=O)Cl)CCCC(=O)Cl
239.14
239.14
Technical
111-19-3
White to Beige
220.0°C (75.0 mmHg)
Authentic
Glass bottle
1.4670 to 1.4700
MFCD00000770
02, 719
sebacoyl chloride, sebacyl chloride, sebacoyl dichloride, sebacic acid dichloride, decanedioyl chloride, sebacoylchloride, octane-1,8-dicarbonyl chloride, sebacoyl chloride, cp, acmc-1byrk, decanedioic acid chloride
WMPOZLHMGVKUEJ-UHFFFAOYSA-N
decanedioyl dichloride
66072
92%
Liquid

Identifiants chimiques

111-19-3
239.14
WMPOZLHMGVKUEJ-UHFFFAOYSA-N
66072
C(CCCCC(=O)Cl)CCCC(=O)Cl
C10H16Cl2O2
MFCD00000770
sebacoyl chloride, sebacyl chloride, sebacoyl dichloride, sebacic acid dichloride, decanedioyl chloride, sebacoylchloride, octane-1,8-dicarbonyl chloride, sebacoyl chloride, cp, acmc-1byrk, decanedioic acid chloride
decanedioyl dichloride
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
Harmful if swallowed.
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
May be corrosive to metals.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breath

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 203-843-4

missing translation for 'rtecsNumber' : HD8454250

missing translation for 'tsca' : TSCA

Recommended Storage : Product may develop some turbidity or precipitate

FDS Documents

Documents

Offres spéciales

Offres spéciales

RUO – Research Use Only