missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sebacoyl chloride, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1422614

 View more versions of this product

Catalog No. AAA1422614


Explore available promotions

Description

Description

Sebacoyl chloride is used in the synthesis of nylon-6,10 polymer by reacting with hexamethylenediamine. It is also used to prepare decanedioic acid bis-(3-dimethylamino-propyl) ester in the presence of triethylamine as catalyst. It is used as an intermediate in chemical synthesis. It is involved in the production of polyamide microcapsules by polycondensation of p-phenylenediamine with sebacoyl chloride using diethylenetriamine (DETA) cross-linker and oleic acid as a plasticizer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sebacoyl chloride is used in the synthesis of nylon-6,10 polymer by reacting with hexamethylenediamine. It is also used to prepare decanedioic acid bis-(3-dimethylamino-propyl) ester in the presence of triethylamine as catalyst. It is used as an intermediate in chemical synthesis. It is involved in the production of polyamide microcapsules by polycondensation of p-phenylenediamine with sebacoyl chloride using diethylenetriamine (DETA) cross-linker and oleic acid as a plasticizer.

Solubility
Miscible with hydrocarbons and ethers.

Notes
Store in a cool place. Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with water, acids, bases, amines, alcohols and oxidizing agents.
Specifications

Specifications

Sebacoyl chloride
-5°C
161°C (14 mmHg)
Acrid
1.468
25 g
1365665
sebacoyl chloride, sebacyl chloride, sebacoyl dichloride, sebacic acid dichloride, decanedioyl chloride, sebacoylchloride, octane-1,8-dicarbonyl chloride, sebacoyl chloride, cp, acmc-1byrk, decanedioic acid chloride
WMPOZLHMGVKUEJ-UHFFFAOYSA-N
decanedioyl dichloride
66072
97%
111-19-3
1.121
165°C (329°F)
C10H16Cl2O2
MFCD00000770
UN2922
Moisture sensitive
Miscible with hydrocarbons and ethers.
C(CCCCC(=O)Cl)CCCC(=O)Cl
239.136
239.14

Chemical Identifiers

111-19-3
239.136
WMPOZLHMGVKUEJ-UHFFFAOYSA-N
66072
C(CCCCC(=O)Cl)CCCC(=O)Cl
C10H16Cl2O2
MFCD00000770
sebacoyl chloride, sebacyl chloride, sebacoyl dichloride, sebacic acid dichloride, decanedioyl chloride, sebacoylchloride, octane-1,8-dicarbonyl chloride, sebacoyl chloride, cp, acmc-1byrk, decanedioic acid chloride
decanedioyl dichloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H310-H314-H318-H302
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

P260-P262-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H310-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

EINECSNumber : 203-843-4

RTECSNumber : HD8454250

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only