missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

sec-Butanol, 99+%, for analysis, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 220290010

 View more versions of this product

Catalog No. AC220290010



Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

sec-Butanol
For analysis
0.8100g/mL
1 L
Glass Bottle
99+%
C4H10O
MFCD00004569
0.1% max.
15, 1543
Solubility in water: 125g/L (20°C). Other solubilities: soluble in alcohol, ester, ether and benzene
CCC(C)O
74.12
CHEBI:35687
≥99%
-115.0°C
78-92-2
99.0°C
24°C
Liquid
0.81
C2H5CH(OH)CH3
4.2 mPa.s (20°C)
01, 371
2-butanol, sec-butanol, sec-butyl alcohol, 2-hydroxybutane, 2-butyl alcohol, s-butyl alcohol, butylene hydrate, s-butanol, 1-methyl-1-propanol, 1-methylpropyl alcohol
BTANRVKWQNVYAZ-UHFFFAOYNA-N
butan-2-ol
6568
74.12
Analytical

Chemical Identifiers

78-92-2
74.12
BTANRVKWQNVYAZ-UHFFFAOYNA-N
6568
butan-2-ol
C4H10O
MFCD00004569
2-butanol, sec-butanol, sec-butyl alcohol, 2-hydroxybutane, 2-butyl alcohol, s-butyl alcohol, butylene hydrate, s-butanol, 1-methyl-1-propanol, 1-methylpropyl alcohol
CHEBI:35687
CCC(C)O
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause drowsiness or dizziness.
Flammable liquid and vapor.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only