Learn More
sec-Butyl acetate, 98%, Thermo Scientific Chemicals
$60.10 - $147.55
Chemical Identifiers
CAS | 105-46-4 |
---|---|
Molecular Formula | C6H12O2 |
Molecular Weight (g/mol) | 116.16 |
MDL Number | MFCD00009328 |
InChI Key | DCKVNWZUADLDEH-UHFFFAOYNA-N |
Synonym | sec-butyl acetate, dl-sec-butyl acetate, 2-butyl acetate, acetic acid, 1-methylpropyl ester, 1-methylpropyl acetate, sec-butyl ethanoate, 1-methylpropyl ethanoate, acetic acid, sec-butyl ester, 2-butanol acetate, sec-butyl alcohol acetate |
PubChem CID | 7758 |
IUPAC Name | butan-2-yl acetate |
SMILES | CCC(C)OC(C)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAL1389214
|
Thermo Scientific Chemicals
L1389214 |
25 g |
Each for $60.10
|
|
||||||
AAL1389222
|
Thermo Scientific Chemicals
L1389222 |
100 g |
Each for $147.55
|
|
||||||
Description
sec-Butyl acetate is used in organic synthesis, catalytic agent, petrochemical additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationssec-Butyl acetate is used in organic synthesis, catalytic agent, petrochemical additive.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
Chemical Identifiers
105-46-4 | |
116.16 | |
DCKVNWZUADLDEH-UHFFFAOYNA-N | |
7758 | |
CCC(C)OC(C)=O |
C6H12O2 | |
MFCD00009328 | |
sec-butyl acetate, dl-sec-butyl acetate, 2-butyl acetate, acetic acid, 1-methylpropyl ester, 1-methylpropyl acetate, sec-butyl ethanoate, 1-methylpropyl ethanoate, acetic acid, sec-butyl ester, 2-butanol acetate, sec-butyl alcohol acetate | |
butan-2-yl acetate |
Specifications
105-46-4 | |
0.872 | |
16°C (60°F) | |
1.389 | |
25 g | |
1720689 | |
sec-butyl acetate, dl-sec-butyl acetate, 2-butyl acetate, acetic acid, 1-methylpropyl ester, 1-methylpropyl acetate, sec-butyl ethanoate, 1-methylpropyl ethanoate, acetic acid, sec-butyl ester, 2-butanol acetate, sec-butyl alcohol acetate | |
DCKVNWZUADLDEH-UHFFFAOYNA-N | |
butan-2-yl acetate | |
7758 | |
98% |
-99°C | |
111°C to 112°C | |
C6H12O2 | |
MFCD00009328 | |
UN1123 | |
14,1536 | |
Slightly soluble in water. | |
CCC(C)OC(C)=O | |
116.16 | |
116.16 | |
sec-Butyl acetate |
Safety and Handling
GHS H Statement
H225
Highly flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H225
DOTInformation : Hazard Class: 3; Packaging Group: II
EINECSNumber : 203-300-1
RTECSNumber : AF7380000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only