missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Epredia™ Section-Drying Hotplate
Designed to dry paraffin tissue sections efficiently; a matte black heating surface coating provides optimal visual contrast for tissue sections.
$9251.70 - $9251.70
Catalog Number | Mfr. No. | Type | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Price | Quantity | |||||
A84600031
|
Epredia™
A84600031 |
Slimline Hotplate |
Each for $9,251.70
|
|
|||||
Description
- Ultra-fast heating system
- Bright LED display
- Anti-microbial coating
- Mini Version: fixed heat setting of 75°C
- Slimline Version: adjustable slide support rails, variable temperature control, and memory function
Specifications
110 V | |
110 V, 50/60 Hz |