Polymedco™ Sedimat™ 15 Plus Sedrate System DFS Item
Fast, in-office results for one to eight specimens simultaneously.
Supplier:  Polymedco™ HT100
 View more versions of this product
Catalog No. 13-641-587B
Explore available promotions
Description
Description
Automated sedrate system provides accurate ESR results in only 15 minutes.
  • Random access capability
  • Faster throughput; processes 32 patient results per hour
  • Eight-position reader
  • Small footprint suitable for tight benchtops
  • Bar code reader available
  • LIS compatible for data transfer
  • Printer available (21-000-123)
Specifications
Specifications
Thermal Printer Paper
Fast, in-office results for one to eight specimens simultaneously
Sedimat™ 15 plus sedrate system
Product Suggestions
Product Suggestions