missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
SEKISUI Diagnostics™ OSOM™ Trichomonas Rapid Test
The OSOM™ Trichomonas Rapid Test is the only CLIA-waived lateral flow rapid test for the detection of Trichomoniasis while providing 95% agreement with culture and wet mount combined.
$118.76 - $881.94
Specifications
Content And Storage | Room temperature (15°C to 30°C, 59°F to 86°F) |
---|---|
CPT Code | 87808QW |
Clia Complexity | Waived |
Test Time | 10 min. |
Analysis Time | 10 min. |
Catalog Number | Mfr. No. | Type | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Price | Quantity | |||||
22-071-072
|
SEKISUI Diagnostics™
SEK182 |
Control Kit |
Pack of 10 Each for $118.76
|
|
|||||
22-071-071
|
SEKISUI Diagnostics™
SEK181 |
Rapid Test |
Pack of 25 Each for $881.94
Case of 6 Pack for $5,291.64
|
|
|||||
Description
- Simple-to-use - get easy to read results in just ten minutes and four easy steps
- Eliminate the need for wet mount - antigen-based rapid test does not require visualization of live organisms
- CDC-approved method of testing for the detection of T. vaginalis - unaffected by other common infections
- Trust your results - 95% agreement against Composite Reference Standard (culture and wet mount)
- Easy-to-use - requires minimal training and no additional equipment
- QC Inside™ - two extra tests included at no additional charge to run external quality control testing for more confidence in your results
- Made in the USA
Specifications
Room temperature (15°C to 30°C, 59°F to 86°F) | |
Waived | |
10 min. |
87808QW | |
10 min. |