missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
SEKISUI Diagnostics™ OSOM™ Ultra Strep A Test
The OSOM™ Ultra Strep A Test offers faster results for faster treatment—statistically equivalent to single swab culture with results in just five minutes.
$263.24 - $511.00
Specifications
Accuracy | 92.6% |
---|---|
Content And Storage | Room temperature (15°C to 30°C, 59°F to 86°F) |
Control Sets | Yes, included |
CPT Code | 87880QW |
Clia Complexity | Waived |
Includes: Controls
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
22071067
|
SEKISUI Diagnostics™
SEK147 |
25 Tests |
Pack of 25 Each for $263.24
Case of 18 Pack for $4,738.32
|
|
|||||
22071068
|
SEKISUI Diagnostics™
SEK149 |
50 Tests |
Pack of 50 Each for $511.00
Case of 18 Pack for $9,198.00
|
|
|||||
Description
- Simple-to-use - get easy to read results in four easy steps and no need for additional equipment
- Greater sensitivity - antibody labeled color particles coated at two separate locations allows for greater sensitivity than conventional single label technology without sacrificing specificity
- Easy-to-read two-color results
- Simplify reagent addition - single reagent delivery system in 25-test kit eliminates the need to handle two separate reagents
- QC Inside™ - two extra tests included in the 50-test kit at no additional charge to run external quality control for more confidence in your results
- Made in the USA
Specifications
92.6% | |
Yes, included | |
Waived | |
No | |
96.4% (95% CI: 93.0% to 97.9%) | |
Test Kit | |
5 min. |
Room temperature (15°C to 30°C, 59°F to 86°F) | |
87880QW | |
Streptococcal Antigen (Group A) | |
Throat Swab | |
Rapid Test | |
96.3% (95% CI: 94.3% to 97.5%) | |
5 min. |