SEKISUI Diagnostics™ OSOM™ H. pylori Test
Use the OSOM™ H. pylori Test to detect Anti-H. pylori IgG antibodies in just three steps and 10 minutes.
$641.15 - $641.15
Specifications
CPT Code 86318QW (Whole Blood), 86677 (Serum, Plasma)
Detectable Analytes H. pylori
Sample Type Whole Blood, Serum, or Plasma
Sensitivity 95.9%
Test Time 10 min.
View More Specs
Products 1
Catalog Number Mfr. No. Type Price Quantity  
Catalog Number Mfr. No. Type Price Quantity  
22071201
View Documents
SEKISUI Diagnostics™
SEK175
Rapid Test
Pack of 25 Each for $641.15
Case of 6 Pack for $3,846.90
Description
Description
  • Simple-to-use - get easy to read results with one reagent and three easy steps, with no need for additional equipment
  • Minimal sample required - test requires 10 μL of serum or plasma; 25 μL of whole blood
  • Trust your results - sensitivity of 95.9% versus biopsy/histology
  • Easy-to-read - two colored lines at both the test (T) and control (C) positions indicate a positive result
Specifications
Specifications
86318QW (Whole Blood), 86677 (Serum, Plasma)
Whole Blood, Serum, or Plasma
10 min.
H. pylori
95.9%
10 min.
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'