missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Selectophore™ Lithium Ionophore VI, Function Tested, 99.0% (HPLC), MilliporeSigma™ Supelco™
$172.26 - $935.27
Chemical Identifiers
CAS | 106868-21-7 |
---|---|
Molecular Formula | C24H32O4 |
Molecular Weight (g/mol) | 384.52 |
MDL Number | MFCD00037498 |
InChI Key | KDMUFHBMXZLDLI-UHFFFAOYSA-N |
Synonym | 6,6-Dibenzyl-1,4,8-11-tetraoxacyclotetradecane; 6,6-Dibenzyl-14-crown-4 |
IUPAC Name | 6,6-dibenzyl-1,4,8,11-tetraoxacyclotetradecane |
SMILES | C(C1=CC=CC=C1)C1(CC2=CC=CC=C2)COCCOCCCOCCOC1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
111012401
|
Merck Emd Millipore
6256710MG |
10 mg | Bottomless Glass Bottle with Fused Cone |
Each for $172.26
|
|
||||
111012402
|
Merck Emd Millipore
62567100MG |
100 mg | Bottomless Glass Bottle with Fused Cone |
Each for $935.27
|
|
||||
Chemical Identifiers
106868-21-7 | |
384.52 | |
KDMUFHBMXZLDLI-UHFFFAOYSA-N | |
6,6-dibenzyl-1,4,8,11-tetraoxacyclotetradecane |
C24H32O4 | |
MFCD00037498 | |
6,6-Dibenzyl-1,4,8-11-tetraoxacyclotetradecane; 6,6-Dibenzyl-14-crown-4 | |
C(C1=CC=CC=C1)C1(CC2=CC=CC=C2)COCCOCCCOCCOC1 |
Specifications
106868-21-7 | |
C24H32O4 | |
MFCD00037498 | |
6,6-Dibenzyl-1,4,8-11-tetraoxacyclotetradecane; 6,6-Dibenzyl-14-crown-4 | |
C(C1=CC=CC=C1)C1(CC2=CC=CC=C2)COCCOCCCOCCOC1 | |
384.52 | |
≥99.0% (HPLC) | |
Selectophore |
Bottomless Glass Bottle with Fused Cone | |
C24H32O4 | |
10 mg | |
KDMUFHBMXZLDLI-UHFFFAOYSA-N | |
6,6-dibenzyl-1,4,8,11-tetraoxacyclotetradecane | |
384.51 | |
Selectophore™ |
Selectophore is a trademark of Sigma-Aldrich Chemie GmbH