missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Selenium(I) chloride, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 0402396Y

 View more versions of this product

Catalog No. AA402396Y


Explore available promotions

Description

Description

Selenium monochloride is used in electronics, instrumentation and other industrial purposes. It is used as a reagent for the synthesis of Se-containing compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

127°C (decomposition)
-85°C
Liquid
Cl2Se2
UN2922
diselenium dichloride, selenium monochloride, selenium chloride se2cl2, unii-2a5292fc4p, selenium i chloride, dichlorodiselenane, dichloro perselenide, se2cl2, selenium i chloride, ampuled under argon trace metals basis 25g
VIEXQFHKRAHTQS-UHFFFAOYSA-N
chloroselanyl selenohypochlorite
66206
99%
Moisture sensitive
Selenium(I) chloride
10025-68-0
2 x 25 g
MFCD00011226
14,8432
Decomposes in water; Soluble in chloroform, benzene, carbon tetrachloride, carbon disulfide
Cl[Se][Se]Cl
228.84
228.83
Acidic
2.77 g/mL

Chemical Identifiers

10025-68-0
228.84
VIEXQFHKRAHTQS-UHFFFAOYSA-N
66206
Cl[Se][Se]Cl
Cl2Se2
MFCD00011226
diselenium dichloride, selenium monochloride, selenium chloride se2cl2, unii-2a5292fc4p, selenium i chloride, dichlorodiselenane, dichloro perselenide, se2cl2, selenium i chloride, ampuled under argon trace metals basis 25g
chloroselanyl selenohypochlorite
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H301+H331-H314-H335-H373

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

EINECSNumber : 233-037-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only