Learn More
Selenium(IV) chloride, 99.5% (metals basis), Thermo Scientific Chemicals
$224.62 - $754.21
Chemical Identifiers
CAS | 10026-03-6 |
---|---|
Molecular Formula | Cl4Se |
Molecular Weight (g/mol) | 220.771 |
MDL Number | MFCD00011225 |
InChI Key | LNBXMNQCXXEHFT-UHFFFAOYSA-N |
Synonym | selenium tetrachloride, selenium chloride secl4 , t-4, unii-4gb8868p5j, selenium iv chloride 1:4, secl4, tetrachloro-, e4-selane, selenium iv chloride, selenium iv chloride-8 mesh, selenium iv chloride trace metals basis 25g |
PubChem CID | 66205 |
SMILES | Cl[Se](Cl)(Cl)Cl |
Description
It is used in the synthesis of other selenium compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10026-03-6 | |
220.771 | |
LNBXMNQCXXEHFT-UHFFFAOYSA-N | |
66205 |
Cl4Se | |
MFCD00011225 | |
selenium tetrachloride, selenium chloride secl4 , t-4, unii-4gb8868p5j, selenium iv chloride 1:4, secl4, tetrachloro-, e4-selane, selenium iv chloride, selenium iv chloride-8 mesh, selenium iv chloride trace metals basis 25g | |
Cl[Se](Cl)(Cl)Cl |
Specifications
(sublimation) | |
Lump | |
Cl4Se | |
UN2923 | |
selenium tetrachloride, selenium chloride secl4 , t-4, unii-4gb8868p5j, selenium iv chloride 1:4, secl4, tetrachloro-, e4-selane, selenium iv chloride, selenium iv chloride-8 mesh, selenium iv chloride trace metals basis 25g | |
LNBXMNQCXXEHFT-UHFFFAOYSA-N | |
220.771 | |
220.77 | |
Pungent | |
Moisture sensitive | |
Selenium(IV) chloride |
10026-03-6 | |
25 g | |
MFCD00011225 | |
14,8438 | |
Decomposes in water and moist air; Insoluble in liquid bromine; Sublimes when heated | |
Cl[Se](Cl)(Cl)Cl | |
66205 | |
99.5% | |
1.807 | |
2.600 g/mL | |
(metals basis) |
Safety and Handling
GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c
H301+H331-H373
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 233-053-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only