Learn More
Brady™ Self-laminating Cryogenic Polyester Labels
Securely label lab tubes and vials for cryogenic storage with this self-laminating, ultra-thin polyester label material (B-461). Print almost anywhere with a compact Brady™ portable label printer.
$61.08 - $95.81
Specifications
| Autoclavable | Autoclavable |
|---|---|
| Adhesive Type | Permanent Acrylic |
| Material | Polyester |
| Product Type | Label |
| Temperature Range (English) | -321°F to +230°F |
| Catalog Number | Mfr. No. | Color | Dimensions (LxW) | For Use With (Equipment) | Format | Price | Quantity | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Color | Dimensions (LxW) | For Use With (Equipment) | Format | Price | Quantity | ||
|
17-012-248
|
Brady™
M5123461 |
White, Transparent | 28.58 mm (W) x 9.53 mm (H) | BMP51;BMP53;M511;M510 | Die-cut |
Each for $83.83
|
|
||
|
17-012-264
|
Brady™
M5122461 |
White, Transparent | 28.58 mm (W) x 12.70 mm (H) | BMP51;BMP53;M511;M510 | Die-cut |
Each for $79.05
|
|
||
|
17-012-226
|
Brady™
M4124461 |
White, Transparent | 41.91 mm (W) x 12.70 mm (H) | BMP41;BMP51;BMP53;M511;M410;M510 | Die-cut |
Each for $77.84
|
|
||
|
17-012-326
|
Brady™
M472461 |
White, Transparent | 44.45 mm (W) x 12.70 mm (H) | BMP41;BMP51;BMP53;M511;M410;M510 | Die-cut |
Each for $74.25
|
|
||
|
17-012-123
|
Brady™
M574461 |
White, Transparent | 0.5 x 2.2 in. | BMP41, BMP51, M511 | Die-cut |
Each for $80.25
|
|
||
|
17-012-246
|
Brady™
M5126461 |
White, Transparent | 0.6 x 1.8 in. | BMP41, BMP51, M511 | Die-cut |
Each for $77.84
|
|
||
|
17-012-308
|
Brady™
M486461 |
White, Transparent | 66.68 mm (W) x 15.24 mm (H) | BMP41;BMP51;BMP53;M511;M410;M510 | Die-cut |
Each for $77.84
|
|
||
|
17-012-178
|
Brady™
M5125461 |
White, Transparent | 44.45 mm (W) x 25.40 mm (H) | BMP51;BMP53;M511;M510 | Die-cut |
Each for $77.84
|
|
||
|
17-012-266
|
Brady™
M575461 |
White, Transparent | 66.68 mm (W) x 25.40 mm (H) | BMP51;BMP53;M511;M510 | Die-cut |
Each for $81.45
|
|
||
|
17-012-255
|
Brady™
M5129461 |
White, Transparent | 57.15 mm (W) x 38.10 mm (H) | BMP51;BMP53;M511;M510 | Die-cut |
Each for $95.81
|
|
||
|
17-012-306
|
Brady™
M5119461 |
White, Transparent | 95.25 mm (W) x 38.10 mm (H) | BMP51;BMP53;M511;M510 | Die-cut |
Each for $61.08
|
|
||
|
17-012-309
|
Brady™
M4C375461 |
White | 9.53 mm (W) x 7.62 m (L) | BMP41;BMP51;BMP53;M511;M410;M510 | Continuous |
Each for $71.86
|
|
||
|
17-012-287
|
Brady™
M4C500461 |
White | 12.70 mm (W) x 7.62 m (L) | BMP41;BMP51;BMP53;M511;M410;M510 | Continuous |
Each for $70.66
|
|
||
|
17-012-116
|
Brady™
M5C1000461 |
White | 25.40 mm (W) x 7.62 m (L) | BMP51;BMP53;M511;M510 | Continuous |
Each for $74.25
|
|
||
|
17-012-207
|
Brady™
M5C1250461 |
White | 31.75 mm (W) x 7.62 m (L) | BMP51;BMP53;M511;M510 | Continuous |
Each for $77.84
|
|
||
Description
- Ideal for curved surfaces such as vials, centrifuge tubes, test tubes, straws and slides
- Able to withstand extreme temperatures from -196°C to 130°C (-320°F to 266°F), immersion in liquid nitrogen and autoclave use
- Resistant to ethanol, toluene, isopropanol, xylene, DMSO, 50% acetic acid, 10% hydrochloric acid, 10% Clorox solution
- Made with a clear overlaminate that protects the print and maintains content visibility
Specifications
| Autoclavable | |
| Polyester | |
| -321°F to +230°F |
| Permanent Acrylic | |
| Label |