missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sellstrom™ Fire Blankets

Lightweight and easy to apply

$324.16 - $324.16

Specifications

Color White
Material Fiberglass
Dimensions (L x W) 72 x 60 in. (182 x 152.4 cm)
For Use With (Application) Rescue operations in offices, industries and hospitals
Length (English) 72 in.
View More Specs

Products 1
Catalog Number Mfr. No. Includes Price Quantity  
Catalog Number Mfr. No. Includes Price Quantity  
17-338-2
View Documents
Sellstrom™
S97450
Wall hanging Vinyl pouch with pull tab
Each for $324.16
 
Description

Description

  • Made of fiberglass and hemmed on all four sides with Kevlar™ thread
  • Brass grommets on corners
  • Machine washable
  • 1.5 × 1.8m (5 × 6 ft.)

  • Red, heavy-duty vinyl pouch protects blanket from exposure to UV rays, chemicals, moisture dust, salt air, insects, and temperature extremes

Use for rescue operations in offices, industries and hospitals: either to cover the fire, cutting off the oxygen supply or to wrap around a person whose clothes are on fire

Specifications

Specifications

White
72 x 60 in. (182 x 152.4 cm)
72 in.
60 in.
Fiberglass
Rescue operations in offices, industries and hospitals
182 cm
152.4 cm
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'