ILC Dover Sentinel XL Blower Kit - Rechargeable NIMH Battery with Duffel
Sentinel XL Blower kit is equipped with all components to get started including XL blower, NiMH rechargeable battery, charging cord, flow meter, quick loc belt, plug, and carrying tote.
Supplier: ILC Dover S300012
Description
- Cartridge filters, breathing tube and hoods sold separately
- Sentinel XL powered air-purifying respirator assemblies and systems include parts required to build a full system or to replace existing parts
- Belt-mounted blower distributes ambient air through a filter to provide clean, breathable air to the wearer
- PAPRs allow workers to move freely and unrestricted
- Sentinel XL system parts are compatible with components of the same brand and series
Specifications
PAPR Assembly, No Headtop | |
Sentinel XL Blower, NiMH Rechargeable battery, charging cord, flow meter, Quick Loc Belt, Plug and carrying tote | |
Sentinel XL |