ILC Dover Sentinel XL CBRN System - Rechargeable NIMH Battery with Backpack
Sentinel XL CBRN PAPR system features an adjustable headband and a unique lay flat design to protect the system while making it easier to store in the backpack.
Supplier: ILC Dover S500002
Description
- Comes with Butyl Hood, Sentinel XL Blower, NiMH battery with charging cord, 3 CBRN cartridge filters and a backpack
- Sentinel XL powered air-purifying respirator assemblies and systems include parts required to build a full system or to replace existing parts
- Belt-mounted blower distributes ambient air through a filter to provide clean, breathable air to the wearer
- PAPRs allow workers to move freely and unrestricted
- Sentinel XL system parts are compatible with components of the same brand and series
Specifications
PAPR System with Hood | |
Comes with Butyl Hood, Sentinel XL Blower, NiMH battery with charging cord, 3 CBRN cartridge filters and a backpack | |
S-5000-02 |