Learn More
ILC Dover Sentinel XT Clear PAPR System with S-3112 Hood - Non-Incendive Blower (Blue) System
The Sentinel XT Clear PAPR System is equipped with the S-3112 clear full hood. Attaches to protective clothing with clips.
Supplier: ILC Dover S332001
Description
- The Sentinel Xt PAPR System contains the Sentinel XT/NI Blower system, 33 in. breathing tube, plug, LiPo Rechargeable battery w/ secure latch and charging cradle, Quick Loc Belt and 2 HEPA cartridge filters
- Powered air-purifying respirator assemblies and systems include parts required to build a full system or to replace existing parts
- Belt-mounted blower distributes ambient air through a filter to provide clean, breathable air to the wearer
- PAPRs allow workers to move freely and unrestricted
- System parts are compatible with components of the same brand and series
Specifications
| PAPR System with Hood | |
| Sentinel XT/NI Blower system, 33 in. breathing tube, plug, LiPo Rechargeable battery with secure latch and charging cradle, Quick Loc Belt and 2 HEPA cartridge filters | |
| Sentinel XT |