missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
EKF HemataCHEK™ Hematocrit Control
Ideal for use with HemataSTAT™ and other microhematocrit centrifuges
Supplier: EKF 300104
Description
- Assayed for HemataSTAT II and other microhematocrit centrifuges
- Comes in Low, Normal and High levels
Order Info
Box of 6 vials; 2.5mL per vial
Specifications
40°C to 80°C | |
Microhematocrit centrifuge | |
For monitoring the Hematocrit determination | |
6 Pack | |
31 Days Open Vial |
Vial | |
HemataCHEK™ Hematology Reference Control | |
Normal, High | |
Human Blood |