Learn More
Thermo Scientific™ Septa
Use low bleed, robust BTO Septa for demanding GC-MS applications.
$100.68 - $333.85
Specifications
For Use With (Equipment) | Trace and Focus GC Instruments |
---|---|
Material | Silicone |
Catalog Number | Mfr. No. | Type | Diameter (Metric) Inner | Description | Quantity | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Diameter (Metric) Inner | Description | Quantity | Price | Quantity | ||
31303211
|
Thermo Scientific™
31303211 |
Low Bleed Septum | 17 mm | BTO; 17 mm | 50 (Glass Jar) |
Pack of 50 Each for $215.19
|
|
||
03-050-304
|
Thermo Scientific™
31303228 |
Low Bleed Septum | 12.7 mm | BTO; 12.7 mm dia.; Max. Temp. 400°C | 50 (Glass Jar) |
Pack of 50 Each for $190.67
|
|
||
31303230
|
Thermo Scientific™
31303230 |
Low Bleed Septum | 11.5 mm | BTO; 11.5 mm dia.; Max. Temp. 400°C | 50 (Glass Jar) |
Pack of 50 Each for $265.89
|
|
||
03-251-650
|
Thermo Scientific™
313G3211 |
Low Coring Septum | 17 mm | TR-Green; 17 mm dia.; Max. Temp. 350°C | 50 (Glass Jar) |
Pack of 50 Each for $191.66
|
|
||
03-251-651
|
Thermo Scientific™
313G3228 |
Low Coring Septum | 12.7 mm | TR-Green; 12.7 mm dia.; Max. Temp. 350°C | 50 (Glass Jar) |
Pack of 50 Each for $185.00
|
|
||
03-251-652
|
Thermo Scientific™
313G3230 |
Low Coring Septum | 11 mm | TR-Green; 11 mm dia.; Max. Temp. 350°C | 50 (Glass Jar) |
Pack of 50 Each for $217.38
|
|
||
313G3240
|
Thermo Scientific™
313G3240 |
Low Coring Septum | 9 mm | TR-Green; 9 mm dia.; Max. Temp. 350°C | 50 (Glass Jar) |
Pack of 25 Each for $175.84
|
|
||
31303240
|
Thermo Scientific™
31303240 |
Low Bleed Septum | 9 mm | BTO Septa; 9 mm; Max. Temp. 400°C | 50 (Glass Jar) |
Pack of 50 Each for $265.89
|
|
||
03-050-320
|
Thermo Scientific™
31303215BP |
Low Coring Septum | 17 mm | BTO; 17 mm dia.; Max. Temp. 400°C | 48 (Blister Pk.) |
Pack of 48 Each for $240.91
|
|
||
03-050-322
|
Thermo Scientific™
31303230BP |
Low Coring Septum | 11.5 mm | BTO; 11.5 mm dia.; Max. Temp. 400°C | 50 (Blister Pk.) |
Pack of 50 Each for $287.04
|
|
||
03-050-323
|
Thermo Scientific™
31303233BP |
Low Coring Septum | 11 mm | BTO; 11 mm dia.; Max. Temp. 400°C | 50 (Blister Pk.) |
Pack of 50 Each for $252.04
|
|
||
03-050-325
|
Thermo Scientific™
31303240BP |
Low Coring Septum | 9 mm | BTO; 9 mm dia.; Max. Temp. 400°C | 50 (Blister Pk.) |
Pack of 50 Each for $290.38
|
|
||
03-050-321
|
Thermo Scientific™
31303250BP |
Low Coring Septum | 12.7 mm | BTO; 12.7 mm dia.; Max. Temp. 400°C | 48 (Blister Pk.) |
Pack of 48 Each for $303.08
|
|
||
03-100-024
|
Thermo Scientific™
313B3233 |
Low Coring Septum | 11 mm | TR-Blue; 11 mm dia.; Max. Temp. 200°C to 250°C | 50 (Glass Jar) |
Pack of 50 Each for $100.68
|
|
||
03-100-023
|
Thermo Scientific™
313B3240 |
Low Coring Septum | 9 mm | TR-Blue; 9 mm dia.; Max. Temp. 200°C to 250°C | 50 (Glass Jar) |
Pack of 50 Each for $100.68
|
|
||
03-050-326
|
Thermo Scientific™
313G3215BP |
Low Coring Septum | 17 mm | TR-Green; 17 mm dia.; Max. Temp. 350°C | 48 (Blister Pk.) |
Pack of 48 Each for $224.08
|
|
||
03-050-328
|
Thermo Scientific™
313G3230BP |
Low Coring Septum | 11 mm | TR-Green; 11 mm dia.; Max. Temp. 350°C | 50 (Blister Pk.) |
Pack of 50 Each for $238.30
|
|
||
03-100-022
|
Thermo Scientific™
313P3211 |
Low Coring Septum | 17 mm | Marathon; 17 mm dia.; Max. Temp. 350°C | 50 (Glass Jar) |
Pack of 50 Each for $262.30
|
|
||
03-050-330
|
Thermo Scientific™
313P3215BP |
Low Coring Septum | 17 mm | Marathon; 17 mm dia.; Max. Temp. 350°C | 48 (Blister Pk.) |
Pack of 48 Each for $276.51
|
|
||
03-050-332
|
Thermo Scientific™
313P3233BP |
Low Coring Septum | 11 mm | Marathon; 11 mm dia.; Max. Temp. 350°C | 50 (Blister Pk.) |
Pack of 50 Each for $241.89
|
|
||
03-050-333
|
Thermo Scientific™
313P3240BP |
Low Coring Septum | 9 mm | Marathon; 9 mm dia.; Max. Temp. 350°C | 50 (Blister Pk.) |
Pack of 50 Each for $333.85
|
|
||
03-050-329
|
Thermo Scientific™
313G3240BP |
Low Coring Septum | 9 mm | TR-Green; 9 mm dia.; Max. Temp. 350°C | 50 (Blister Pk.) |
Pack of 50 Each for $235.92
|
|
||
Description
Ensure optimal performance of your GC instrument with bleed and temperature optimized Thermo Scientific™ BTO and TR-Green Septa. Made of low-bleed silicone, these septa have excellent mechanical properties, are ideal for demanding GC-MS applications, and may be used reliably up to 400 °C.
- Variety of materials and temperature limits for all applications
- For use in Thermo Scientific™ TRACE™ and FOCUS™ GCs and instruments from other manufacturers
- Contaminant-free in individual blister packs
Specifications
Trace and Focus GC Instruments | |
Silicone |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.