missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
STERIS Septihol™ WFI Sterile Alcohol Aerosol Solution
Bag-on-valve aerosol can sprays at any angle to remove residue from hard surfaces and minimize waste; sterile and double bagged for easy introduction into cleanrooms.
Supplier: STERIS 6250E7
Description
- Mixture of 70% (v/v) USP Isopropanol, and 30% USP Water For Injection (WFI)
- Removes residue from hard surfaces, even from the most critical surfaces
- Free of stabilizers and denaturants for quick, and complete evaporation
- Bag-on-valve aerosol design eliminates contact between product and pressurizing nitrogen, permits operation at any angle, and allows maximum product discharge
- All cans are filled and double bagged in a Grade B (Class 100) facility
- Filtered to 0.2 micron
- Gamma irradiated to ensure sterility for easy introduction in critical areas
- Each lot is documented by Certificates of Analysis that contain Endotoxin, Irradiation and Sterility results
- These products are not registered as disinfectants by the US EPA
Specifications
Alcohol Solution | |
70% (v/v) USP Isopropanol Alcohol, 30% USP Water for Injection (WFI) | |
Aerosol | |
24/Cs. | |
300 mL |
Sterile | |
Filtered | |
Double Bagged in a Grade B (Class 100) Facility | |
10.1 oz. |