Applied Biosystems™ SeqStudio™ Genetic Analyzer System with SmartStart
The Applied Biosystems SeqStudio Genetic Analyzer is a 4-capillary, fluorescence-based capillary electrophoresis system designed to provide users with an integrated experience that is easy, efficient, and versatile for research in genetic analysis.
Supplier: Applied Biosystems™ A35645
Description
- The Applied Biosystems SeqStudio Genetic Analyzer is a 4-capillary, fluorescence-based capillary electrophoresis system designed to provide users with an integrated experience that is easy, efficient, and versatile for research in genetic analysis
- Combining an extended warranty with your system purchase, Help extend the life of your investment. These packages include the system, SmartStart, and an extended warranty.
- Simplified instrument software with interactive touch screen for greater ease of use
- Minimized hands-on setup time with integrated reagents cartridge (SeqStudio Cartridge)
- Application flexibility to run Sanger sequencing and fragment analysis on the same run
- Connectivity: Wi-Fi or wired Ethernet connection
- Web-enabled remote monitoring capability
- Consumables usage tracking through radio frequency identification (RFID)
- SmartStart is performed by a Field Application Scientist (FAS) in your lab. SmartStart helps you:
- Get up and running quickly
- Get connected to your local network and Cloud
- Choose appropriate reagents and consumables to meet your needs
- Content will vary depending on your level of knowledge and experience.
Specifications
SeqStudio System + SmartStart + 1 year extended warranty | |
1 Year Extended Warranty | |
Room Temperature |
SeqStudio | |
System + SmartStart + 1 yr ext warranty |
For Research Use Only. Not for use in diagnostic procedures.