MilliporeSigma™ SeQuant™ ZIC™-HILIC Analytical PEEK Columns
PEEK coated HPLC column
Supplier: MilliporeSigma™ 1504400001
Description
SeQuant™ ZIC™-HILIC is the ideal choice for separation of polar and hydrophilic compounds. The high-performance, truly zwitterionic stationary phase, ensures reproductible retention of all types of compounds that have poor retention and are difficult to separate on reversed-phase HPLC columns. ZIC™-HILIC is designed for compounds such as acids and bases, anions and cations, carbohydrates, metabolites, metal complexes, amino acids, peptides, protein digests and much moreSpecifications
2.1 mm | |
3.5 μm | |
100 AÌ | |
Metal-free PEEK |
50 mm | |
100 Å | |
1/Pk. | |
Column |