MilliporeSigma™ SeQuant™ ZIC™-HILIC Analytical PEEK Columns
PEEK coated HPLC column
Supplier:  MilliporeSigma™ 1504460001
 View more versions of this product
Catalog No. M1504460001
Explore available promotions
Description
Description
SeQuant™ ZIC™-HILIC is the ideal choice for separation of polar and hydrophilic compounds. The high-performance, truly zwitterionic stationary phase, ensures reproductible retention of all types of compounds that have poor retention and are difficult to separate on reversed-phase HPLC columns. ZIC™-HILIC is designed for compounds such as acids and bases, anions and cations, carbohydrates, metabolites, metal complexes, amino acids, peptides, protein digests and much more
Specifications
Specifications
4.6 mm
3.5 μm
200 Å
Metal-free PEEK
50 mm
200 Å
1/Pk.
Column