MilliporeSigma™ SeQuant™ ZIC™-HILIC Analytical PEEK Columns
PEEK coated HPLC column
Supplier:  MilliporeSigma™ 1504550001
 View more versions of this product
Catalog No. M1504550001
Explore available promotions
Description
Description
SeQuant™ ZIC™-HILIC is the ideal choice for separation of polar and hydrophilic compounds. The high-performance, truly zwitterionic stationary phase, ensures reproductible retention of all types of compounds that have poor retention and are difficult to separate on reversed-phase HPLC columns. ZIC™-HILIC is designed for compounds such as acids and bases, anions and cations, carbohydrates, metabolites, metal complexes, amino acids, peptides, protein digests and much more
Specifications
Specifications
4.6 mm
5 μm
200 Å
Metal-free PEEK
150 mm
200 Å
1/Pk.
Column