missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Applied Biosystems™ Sequence Detection Primer DFS Item

Sequence Detection Primer

Supplier:  Applied Biosystems™ 4304970

 View more versions of this product

Catalog No. 43-049-70


Explore available promotions

Description

Description

These unlabeled primers can be used with TaqMan™ probes in any real-time PCR application. Choose between dry and in-solution formatted delivery. All primers are desalted.
Specifications

Specifications

7500 System, 7500 Fast System, 7900HT System, StepOne™, StepOnePlus™, ViiA™ 7 System, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio™ 12k Flex
1 tube containing an unlabeled primer in solution or dried down.

Store at -15°C to -25°C
Room Temperature
10,000 pmol
Real Time PCR Applications
Sequencing Primer
Desalted
Unlabeled
Applied Biosystems™
Liquid or Dried Down
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.