Learn More
Thermo Scientific™ Sequential Access Storage Option for the Orbitor™ RS2 Microplate Mover for QuantStudio™ Real-time PCR Systems
Designed for use with the Orbitor RS2 Microplate Mover and a QuantStudio real-time PCR system, enabling real-time PCR in automated environments for increased sample throughput
Supplier: Thermo Scientific™ A43781
Description
- This Sequential Access Storage Option is designed for use with the Orbitor RS2 Microplate Mover and a QuantStudio real-time PCR system, enabling real-time PCR in automated environments for increased sample throughput.
- This storage option holds up to 20 plates/3 stacks (96-well) and up to 40 plates/3 stacks (384-well). These stacks provide the option for the Orbitor RS2 mover to run until empty, which allows for additional plates to be added while the Orbitor RS2 mover is running. This option is ideal for 0.1-mL and 0.2-mL 96-well plates and 384-well plates.
- Couple this Sequential Access Storage Option with the Orbitor RS2 Microplate Mover for QuantStudio Systems to complete your automation bundle.
- For integration of two QuantStudio systems with the Orbitor RS2 mover, the Dual QuantStudio System Integration Kit is required in addition to the automation bundle.
Specifications
| QuantStudio 7 Pro, QuantStudio 7 Flex, or QuantStudio 12 K Flex Real-Time PCR Systems and The Orbitor RS2 Microplate Mover | |
| Room Temperature | |
| 1 Ea. |
| Real Time PCR (qPCR) | |
| Storage Option |
For Research Use Only. Not for use in diagnostic procedures.