missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sera-Mag™ Select

Delivers selective binding of DNA fragments for use with next-generation sequencing (NGS) and PCR clean-up.

$465.33 - $8689.56

Specifications

Content And Storage 2°C to 8°C
For Use With (Application) Tested functionally in size selection
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
09928106
View Documents
Cytiva
29343045
5 mL
Each for $465.33
Only null left
 
09928107
View Documents
Cytiva
29343052
60 mL
Each for $1,733.99
Only null left
 
09928108
View Documents
Cytiva
29343057
450 mL
Each for $8,689.56
Only null left
 
Description

Description

Sera-Mag Select PCR clean-up and size selection reagent combines the exceptional binding characteristics of Sera-Mag Carboxyl Speedbeads with an optimized binding solution in a ready-to-use formulation. The DNA size range isolated from the process can be tailored to suit the user’s end requirements by adjusting the amount of reagent that is mixed with a fixed volume of sample.

  • Use a single reagent for PCR clean-up and size selection
  • Easily control what size DNA is retained or eliminated by adding various ratios of Sera-Mag Select to your sample, enabling NGS library and PCR fragment clean-up, as well as dual size selection of libraries
  • Uses the same protocols as existing reagents so it can be directly integrated into existing workflows with minimal disruption
  • Sera-Mag Select delivers tight distribution in size selection for high yield recovery of specific fragment sizes
  • Suitable for manual, high throughput and automated workflows
  • Sera-Mag Select should be stored in the refrigerator between 2°C to 8°C - Do not freeze - Store upright and keep bottle tightly sealed
Specifications

Specifications

2°C to 8°C
Tested functionally in size selection
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'