missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Sera-Mag SpeedBeads™ Streptavidin magnetic beads
SpeedBeads Streptavidin exhibit a high affinity when binding to biotinylated ligands such as proteins, nucleic acids and peptides. They respond much faster to a magnetic field to shorten assay times and improve precision and also move faster through viscous solutions.
$510.40 - $8960.00
Specifications
| Concentration | 1% solids (10mg/mL) |
|---|---|
| Content And Storage | 2°C to 8°C |
| For Use With (Application) | Immunoassays, cDNA Library Construction, Capture of Biotinylated Targets |
| Diameter (Metric) | 1 μm |
Description
- Available with a 3500-4500 pmol/mg biotin-binding range
- Their “cauliflower-like” structure gives Sera-Mag beads high binding capacity
- Combine fast reaction kinetics and low, non-specific binding for increased throughput and precision in such immunoassay and molecular biology applications as sample preparation and assay development for genomics and proteomics
Specifications
| 1% solids (10mg/mL) | |
| 2°C to 8°C | |
| Immunoassays, cDNA Library Construction, Capture of Biotinylated Targets | |
| 1 μm |