Invitrogen™ Serotonin Competitive ELISA Kit
ELISA
Supplier: Invitrogen™ EEL006
Description
Serotonin (also known as 5-hydroxytryptamne or 5-HT) is a biochemical messenger and neurotransmitter synthesized from L-tryptophan. Serotonin has a range of functions, including regulation of gastric motility, and may be related to some psychiatric disorders including schizophrenia and depression. Serotoinin is widely distributed in the central and peripheral nervous system, and also in some non-nueral tissues such as platelets.Specifications
15.63 to 1000 ng/mL | |
HRP | |
Plasma, Serum | |
<10% | |
Pre-coated 96 well plate, Plate Sealer, Standard, Biotinylated Detection Antibody, Standard & Sample Diluent, Standard & Sample Diluent, HRP Conjugate Diluent, Wash Buffer, Wash Buffer, Stop Solution, Stop Solution | |
96 Tests | |
Plasma 50 μL, Serum 50 μL | |
Serotonin | |
3 hrs 30 mins |
9.38 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
<10 | |
HRP | |
RUO | |
Refer to product documentation for component specific storage temperature. | |
1 hrs 20 mins |