Seward™ Stomacher™ Model 400 Circulator Lab Blender, 110V
Stomacher™ lab blenders offer fast reliable blending and homogenisation of a broad range of laboratory samples. The Stomacher 400 Circulator has a patented paddle action designed to maximize the extraction of micro-organisms from food samples
Supplier:  Seward™ 030010108
Catalog No. 14-285-29
Explore available promotions
Description
Description
Curved shaped paddle produces crushing, washing and circulation of the sample in the bag; Patented curved shaped paddles have been microbiologically proven to out-perform all rectangular paddle models across a broad range of food sample types and textures; One-touch, user-programmable time and speed functions; Three paddle speeds, variable time settings from 1 second to 99 minutes and 59 seconds; Auto-run for hands-free operation
Specifications
Specifications
Circulator
110 V
60 Hz
2.7 to 13.52 oz.
13.25 in.
35.5 x 33 x 30 cm
30 cm
13.25 in.
Ideal for maximize the extraction of microorganisms from food samples
Stomacher 400
300 rpm
110 V, 60 Hz
170 W
80 to 400 mL
35.5 cm
11 in.
35.5 cm
33 cm
Power cables, user manual, stomacher bags, 3 year warranty, 10 years service and spares support
1 Ea.
Videos
SDS
Documents
Documents
Accessories