Epredia™ Cryochrome™ Embedding Resin
Support tissue for frozen sectioning with Epredia™ Cryochrome™ Embedding Resin.
$239.32 - $319.59
Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
28-611-45
|
Epredia™
9990422 |
Embedding Media |
Case of 4 Each for $239.32
|
|
|||||
28-610-95
|
Epredia™
9990426 |
Embedding Media |
Case of 4 Each for $319.59
|
|
|||||
28-611-47
|
Epredia™
9990428 |
Embedding Media |
Case of 4 Each for $263.00
|
|
|||||
Description
- Color codes frozen sections for easy identification and tracking
- Four colors available: pink, blue, yellow, green
- Formulated with physical characteristics identical to Cryomatrix™ embedding resin
Specifications
Embedding Media | |
120mL |