Epredia™ Cryomatrix™ embedding resin
Secure and support tissue with Epredia™ Cryomatrix™ embedding resin, an outstanding product used to support tissue for frozen sectioning.
Supplier: Epredia™ 6769006
Description
- Forms a firm bond with the specimen holder
- Freezes quickly, sections smoothly
- Dissolves in water without residue
Specifications
120 mL | |
Embedding Media |