Epredia™ Cytoclip™ Stainless-Steel Slide Clip
The Epredia™ Cytoclip Stainless-Steel Slide Clip holds Cytofunnel™ disposable sample chamber against the microscope slide.
Supplier:  Epredia™ 59910052
Catalog No. 59-910-052
Explore available promotions
Description
Description
  • For preparation in the Epredia Shandon Cytospin™ Cytocentrifuge
  • Autoclavable and reusable
Specifications
Specifications
Stainless Steel
6/Box
Side Clip
Product Suggestions
Product Suggestions