Epredia™ Cytospin™ Collection Fluid
Store specimens or unstained slides for weeks without adverse effect with Epredia™ Shandon™ Cytospin™ Collection Fluid.
Supplier:  Epredia™ 9990310
 View more versions of this product
Catalog No. 28-601-79
Explore available promotions
Description
Description
  • Packaged in convenient pump bottle or pre-filled specimen cups
  • Ready-to-use alcohol/Carbowax™ (PEG) combination for direct specimen collection and fixation
Specifications
Specifications
Collection Fluid
10L
Alcohol/Carbowax (PEG) combination