Epredia™ Double Cytoslide™ Microscope Slides
Use Epredia™ Shandon™ Double Cytoslides™ with Epredia™ Shandon™ EZ Double Cytofunnel™ Chambers.
Supplier: Epredia™ 5991055
Description
- Specimen is displayed within a circle marked as 1 or 2
- Number circles correspond with the Cytofunnel™ chamber
Ordering Alert:
- Sold in boxes of 100 slides
Specifications
Double Cytoslide | |
Glass | |
Plain | |
Coated |