Epredia™ Shandon™ Harris Hematoxylin
Take advantage of acidified or non-acidified formulations with the Epredia™ Shandon™ Harris Hematoxylin. This product does not employ a mercury-based oxidizing agent, thus removing a possible source of environmental contamination.
Supplier:  Epredia™ 6765002
 View more versions of this product
Catalog No. 28-600-99
Explore available promotions
Description
Description
  • Acidifed: Intensely stained, blue-black nuclei
  • Non-acidified: Intensely stained, blue-black nuclei
Specifications
Specifications
Hematoxylin
4L