missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Epredia™ Shandon-Mount™

Perform quick, high-quality conventional coverslipping with Shandon-Mount™ permanent mounting medium.

Supplier:  Epredia™ 1900331

 View more versions of this product

Catalog No. 28-600-61


Explore available promotions

Description

Description

  • Overnight-drying liquid cover slip
  • Toluene base
  • Refractive index: 1.495 ±0.005 (dry)
  • Viscosity: Low (72-120 cps at 22°C)
Specifications

Specifications

Mounting Medium
60 mL
Videos
SDS
Documents

Documents

Promotions

Promotions