Epredia™ Xylene Substitute
Achieve safer, more convenient processing, staining and coverslipping with Epredia™ Xylene Substitute.
Supplier: Epredia™ 9990507
Description
- Not sensitizing to normal skin
- Evaporation rate similar to xylene
- Virtually odorless aliphatic hydrocarbon—no citrus aroma
- Airborne exposure limit three times safer than xylene (TLV 300ppm)
Specifications
5 gal. |