missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sharps Containers
Designed to fit into blood drawing trays for point-of-use disposal
$9.60 - $29.35
Specifications
Certifications/Compliance | BSE/TSE |
---|---|
Material | Polypropylene |
Product Type | Sharps Container |
Catalog Number | Mfr. No. | Capacity (English) | Dimensions (L x W x H) | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (English) | Dimensions (L x W x H) | Price | Quantity | ||||
12500102
|
Zhejiang Gong Dong
SC01Q |
1 qt. | 10.8 x 8.8 x 17.4 cm (4.3 x 3.5 x 6.9 in.) |
Each for $9.60
Case of 60 Each for $576.00
|
|
||||
12500100
|
Zhejiang Gong Dong
SC05Q |
5 qt. | 27 x 11.6 x 29 cm (10.6 x 4.6 x 11.4 in.) |
Each for $29.35
Case of 20 Each for $587.00
|
|
||||
Description
- Meet the requirements of BSE/TSE
- Manufactured under the ISO 13485 standard
- Puncture-Proof and Durable Construction
- Meets Medical Biohazard Waste Regulations
Specifications
BSE/TSE | |
Polypropylene | |
Sharps Container |