IgM, (Fc) Sheep anti-Human, Polyclonal, Secondary Antibody, MilliporeSigma™
Sheep Polyclonal Antibody
Supplier:  MilliporeSigma AP512
Catalog No. AP512MI
Explore available promotions
Description
Description
Sheep anti-Human IgM, (Fc) Antibody is an antibody against Human IgM, (Fc) for use in ELISA.
Specifications
Specifications
Sheep anti-Human IgM
Polyclonal
Purified sheep polyclonal in PBS containing 0.05% sodium azide. Depleted against rabbit IgM and human IgG.
Human anti-Human IgM (myeloma) Fc5u fragment
500 μg
Human
Purified
ELISA
Unconjugated
Sheep
Depletion
Secondary
2-8°C for up to 12 months.