missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Labconco™ Shelves for FreeZone™ Freeze Dryers
Accommodate bulk samples, microtiter plates, vials, or serum bottles for bulk drying applications.
$2208.31 - $2724.22
Specifications
Material | Stainless Steel |
---|---|
For Use With (Application) | Accommodate bulk samples, microtiter plates, vials, or serum bottles for bulk drying applications |
For Use With (Equipment) | FreeZone™ Drying Chambers |
Width (English) | 10.4 in. |
Length (English) | 10.9 in. |
Catalog Number | Mfr. No. | Includes | Weight (English) | Height (English) | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Weight (English) | Height (English) | Price | Quantity | |||
16-108-571
|
Labconco™
7317200 |
Five aluminum shelves with 1/4 in. rims, one stainless steel support, and three rubber feet | 5.5 lb. | 17.5 in. |
Each for $2,724.22
|
|
|||
16-108-570
|
Labconco™
7317100 |
Three aluminum shelves with 1/4 in. rims, one stainless steel support, and three rubber feet | 3.5 lb. | 9.8 in. |
Each for $2,208.31
|
|
|||
Description
Accommodate bulk samples, microtiter plates, vials, or serum bottles for bulk drying applications.
- Three removable, 10 in. diameter aluminum shelves with 1/4 in. rim
- Shelves slide into stainless steel support
- Includes 3 rubber feet
- Design allows for flexible shelf height spacing
- Shelf can be placed in freezer to prefreeze sample for quick loading
- Designed to fit inside Drying Chambers (sold separately)Also available: Heated shelves that include ambient to +40°C (+104°F) variable heat control on models manufactured 2016 and earlier and up to +60°C (+140°F) on models 2017 and later (contact your Fisher Customer Service Representative for details).
Specifications
Stainless Steel | |
FreeZone™ Drying Chambers | |
10.9 in. |
Accommodate bulk samples, microtiter plates, vials, or serum bottles for bulk drying applications | |
10.4 in. |