missing translation for 'onlineSavingsMsg'
Learn More

(-)-Shikimic acid, 98%

CAS: 138-59-0 | C7H9O5- | 173.144 g/mol

$70.45 - $208.30

Chemical Identifiers

CAS 138-59-0
Molecular Formula C7H9O5-
Molecular Weight (g/mol) 173.144
MDL Number MFCD00066278
InChI Key JXOHGGNKMLTUBP-HSUXUTPPSA-M
Synonym shikimic acid, shikimate, --shikimic acid, l-shikimic acid, 3r,4s,5r-3,4,5-trihydroxycyclohex-1-enecarboxylic acid, 3r,4s,5r-3,4,5-trihydroxycyclohex-1-ene-1-carboxylic acid, --shikimate, ccris 7681, unii-29ms2wi2nu, 3alpha,4alpha,5beta-trihydroxy-1-cyclohexene-1-carboxylic acid
PubChem CID 7057976
ChEBI CHEBI:36208
IUPAC Name (3R,4S,5R)-3,4,5-trihydroxycyclohexene-1-carboxylate
SMILES C1C(C(C(C=C1C(=O)[O-])O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL04848MD
View Documents
Thermo Scientific Chemicals
L04848MD
250 mg
Each for $70.45
Only null left
 
AAL0484803
View Documents
Thermo Scientific Chemicals
L0484803
1 g
Each for $208.30
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Specifications

Chemical Identifiers

138-59-0
173.144
JXOHGGNKMLTUBP-HSUXUTPPSA-M
7057976
(3R,4S,5R)-3,4,5-trihydroxycyclohexene-1-carboxylate
C7H9O5-
MFCD00066278
shikimic acid, shikimate, --shikimic acid, l-shikimic acid, 3r,4s,5r-3,4,5-trihydroxycyclohex-1-enecarboxylic acid, 3r,4s,5r-3,4,5-trihydroxycyclohex-1-ene-1-carboxylic acid, --shikimate, ccris 7681, unii-29ms2wi2nu, 3alpha,4alpha,5beta-trihydroxy-1-cyclohexene-1-carboxylic acid
CHEBI:36208
C1C(C(C(C=C1C(=O)[O-])O)O)O

Specifications

138-59-0
Odorless
MFCD00066278
4782717
14,8480
JXOHGGNKMLTUBP-HSUXUTPPSA-M
C1C(C(C(C=C1C(=O)[O-])O)O)O
173.144
CHEBI:36208
98%
184°C to 188°C
C7H9O5-
250 mg
Hygroscopic
shikimic acid, shikimate, --shikimic acid, l-shikimic acid, 3r,4s,5r-3,4,5-trihydroxycyclohex-1-enecarboxylic acid, 3r,4s,5r-3,4,5-trihydroxycyclohex-1-ene-1-carboxylic acid, --shikimate, ccris 7681, unii-29ms2wi2nu, 3alpha,4alpha,5beta-trihydroxy-1-cyclohexene-1-carboxylic acid
−175° (c=2 in Water)
(3R,4S,5R)-3,4,5-trihydroxycyclohexene-1-carboxylate
7057976
174.16
(-)-Shikimic acid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 205-334-2

RTECSNumber : GW4600000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only