Dräger™ Short-Term Detector Tubes - De Tube Hydrogen Sulfide
For gas detection
Supplier:  Dräger™ 6728821
 View more versions of this product
Catalog No. 17-985-247
Explore available promotions
Description
Description
Note: Extension of measuring range listed is possible for some detector tubes. Call Fisher Customer Service for more information.
Specifications
Specifications
Hydrogen Sulfide 2/a
2 to 200ppm
10
32°F to 104°F
3 to 30mg/L
1/10
3.5 min./20 sec.
0°C to 40°C