Dräger™ Short-Term Detector Tubes - De Tube Hydrogen Sulfide
For gas detection
Supplier:  Dräger™ CH28101
 View more versions of this product
Catalog No. 17-985-250
Explore available promotions
Description
Description
Note: Extension of measuring range listed is possible for some detector tubes. Call Fisher Customer Service for more information.
Specifications
Specifications
Hydrogen Sulfide 0.2%/a
0.2 to 7 vol.%
10
32°F to 140°F
40mg/L
1+2
2 min.
0°C to 60°C