Dräger™ Short-Term Detector Tubes - De Tube Hydrogen Sulfide
For gas detection
Supplier:  Dräger™ CH28201
 View more versions of this product
Catalog No. 17-985-252
Explore available promotions
Description
Description
Note: Extension of measuring range listed is possible for some detector tubes. Call Fisher Customer Service for more information.
Specifications
Specifications
Hydrogen Sulfide, Sulfur Dioxide 0.2%/A
0.2 to 7 vol.%
10
32°F to 104°F
40mg/L
1+2
2 min.
0°C to 40°C